Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122759.2 | NP_001116231.1 | 6 - 7 | 908 | 106 | |
NM_008802.3 | NP_032828.2 | 6 - 7 | 638 | 106 | ||
GenBank mRNA | AK036517.1 | - | 6 - 7 | 596 | 106 | |
AK154521.1 | - | 6 - 7 | 725 | 106 | ||
AK168957.1 | - | 6 - 7 | 600 | 106 | ||
AY007702.1 | - | 6 - 7 | 587 | 106 | ||
BC062909.1 | - | 6 - 7 | 907 | 6815827 | 106 | |
U68171.1 | - | 6 - 7 | 586 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |