Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psen1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008943.2 | NP_032969.1 | 9 - 10 | 1446 | 65 | |
GenBank mRNA | AK134232.1 | - | 9 - 10 | 1129 | 65 | |
AK134615.1 | - | 9 - 10 | 1168 | 65 | ||
AK148101.1 | - | 9 - 10 | 1167 | 65 | ||
AK165984.1 | - | 9 - 10 | 1167 | 65 | ||
AK169963.1 | - | 5 - 6 | 573 | 65 | ||
AK205380.1 | - | 3 - 4 | 218 | 65 | ||
BC030409.1 | - | 9 - 10 | 1121 | 5133302 | 65 | |
BC071233.1 | - | 9 - 10 | 1185 | 3673609 | 65 | |
L42177.1 | - | 9 - 10 | 1142 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |