Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Runx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145920.2 | NP_001139392.1 | 5 - 6 | 2010 | 65 | |
NM_001146038.2 | NP_001139510.1 | 7 - 8 | 1275 | 65 | ||
NM_001271627.1 | NP_001258556.1 | 6 - 7 | 1439 | 65 | ||
NM_001271630.1 | NP_001258559.1 | 4 - 5 | 1364 | 65 | ||
NM_001271631.1 | NP_001258560.1 | 3 - 4 | 1259 | 65 | ||
NM_009820.5 | NP_033950.2 | 7 - 8 | 1242 | 65 | ||
NR_073392.1 | 6 - 7 | 1026 | 65 | |||
GenBank mRNA | AF005936.1 | - | 6 - 7 | 1393 | 65 | |
AF010284.1 | - | 7 - 8 | 1242 | 65 | ||
AF053956.1 | - | 4 - 5 | 554 | 65 | ||
AK089291.1 | - | 3 - 4 | 794 | 65 | ||
D14636.1 | - | 5 - 6 | 2008 | 65 | ||
DQ458792.1 | - | 6 - 7 | 1027 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Inventoried Probe spans exons |