Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011715.2 | NP_035845.1 | 3 - 4 | 291 | 66 | |
GenBank mRNA | AF020055.1 | - | 3 - 4 | 291 | 66 | |
AK004644.1 | - | 3 - 4 | 399 | 66 | ||
AK004858.1 | - | 3 - 4 | 362 | 66 | ||
AK151146.1 | - | 3 - 4 | 290 | 66 | ||
AK156300.1 | - | 3 - 4 | 328 | 66 | ||
AK165225.1 | - | 3 - 4 | 261 | 66 | ||
AK165323.1 | - | 3 - 4 | 261 | 66 | ||
AK167241.1 | - | 3 - 4 | 317 | 66 | ||
AK167275.1 | - | 3 - 4 | 359 | 66 | ||
AK167813.1 | - | 3 - 4 | 289 | 66 | ||
AK169835.1 | - | 3 - 4 | 385 | 66 | ||
AK169893.1 | - | 3 - 4 | 321 | 66 | ||
BC049117.1 | - | 3 - 4 | 398 | 66 | ||
BC119358.1 | - | 3 - 4 | 247 | 8734107 | 66 | |
BC125295.1 | - | 3 - 4 | 247 | 40129810 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |