Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163511.1 | NP_001156983.1 | 6 - 7 | 1342 | 94 | |
NM_021343.2 | NP_067318.2 | 6 - 7 | 1339 | 94 | ||
GenBank mRNA | AF049099.1 | - | 6 - 7 | 1321 | 94 | |
AK011111.2 | - | 6 - 7 | 1306 | 94 | ||
AK144998.1 | - | 6 - 7 | 1669 | 94 | ||
BC145302.1 | - | 6 - 7 | 1310 | 9053856 | 94 | |
BC145309.1 | - | 6 - 7 | 1310 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |