Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vimp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024439.3 | NP_077759.3 | 2 - 3 | 275 | 64 | |
GenBank mRNA | AF335543.1 | - | 2 - 3 | 229 | 64 | |
AK005204.1 | - | 2 - 3 | 258 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |