Assay Details
Gene Symbol: Tmem54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290706.1 | NP_001277635.1 | 4 - 5 | 628 | 80 | |
NM_025452.4 | NP_079728.2 | 4 - 5 | 646 | 80 | ||
NM_133881.1 | NP_598642.1 | 3 - 4 | 586 | 80 | ||
GenBank mRNA | AK007525.1 | - | 3 - 4 | 502 | 80 | |
AK008935.1 | - | 4 - 5 | 561 | 80 | ||
AK009830.1 | - | 3 - 4 | 482 | 80 | ||
BC019563.1 | - | 4 - 5 | 575 | 4219997 | 80 | |
BQ032787.1 | - | 2 - 3 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23294938 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23294937 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23294939 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23294940 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2181888 | BQ032787.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2181888 | NM_025452.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2181888 | NM_133881.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2181888 | AK008935.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2181888 | AK009830.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 745071 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 891149 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 493122 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 63692 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P113888 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2181888 | BQ032787.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2181888 | NM_025452.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2181888 | NM_133881.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2181888 | AK008935.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2181888 | AK009830.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23294952 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23294938 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23294951 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23294919 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23294948 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23294934 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23294932 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23294920 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23294931 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23294913 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23294954 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23294956 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23294942 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23294941 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23294917 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23294922 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23294944 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23294924 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23294959 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23294928 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23294926 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23294937 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23294930 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23294947 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23294949 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23294921 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23294939 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23294940 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23294945 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23294915 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23294957 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P113888 | - | - |
Whole Mouse Genome G4122A | A_51_P303079 | BQ032787.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P303079 | AK007525.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P303079 | NM_025452.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P303079 | NM_133881.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P303079 | AK008935.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140301 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129867 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P113888 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03730179 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03576238 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03730173 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23294940 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23294938 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23294937 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23294939 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294921 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23295001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23295030 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294934 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294930 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23295041 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294954 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23295031 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294998 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23295043 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23295047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294935 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23295035 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294898 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23295020 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294981 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294894 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23295044 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294919 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23295022 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23295049 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294970 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294961 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294959 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294962 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23295006 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23295033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23295029 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23295004 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23295037 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294900 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294945 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294976 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294978 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294902 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294996 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294938 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23295007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294926 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294924 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294899 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23295015 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23295039 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294947 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23295011 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294942 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23295009 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23295034 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294949 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23295025 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23295027 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294905 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294911 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294983 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294920 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294971 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294994 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294966 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23295016 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294907 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23295018 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23295000 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294997 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294910 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294897 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294968 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294917 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23295005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23295026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294928 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294941 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294951 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294956 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23295017 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294915 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294932 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23294995 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03632615 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03486004 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23294972 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03855096 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03730173 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23294910 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23294922 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03850091 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04101060 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03231355 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23294930 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23294920 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03968557 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23294894 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04101058 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03360161 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03695982 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23294935 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03730170 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03730163 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23294976 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03730162 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03943501 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03695985 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23294952 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03486005 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23294978 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23294983 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03855098 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23294899 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23294911 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23294966 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03486006 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03322268 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03943502 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23294977 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23294968 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23294921 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23294961 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03730168 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04101049 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04101057 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03730179 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23294913 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23294945 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04101059 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23294924 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03730164 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04101056 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03231354 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03231349 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03943500 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23294926 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23294975 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10508408 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10508411 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10508410 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10508409 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10508407 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10508406 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1417895_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10508405 | - | - |
Mouse Genome 430 2.0 Array | 1417895_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1417895_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66260_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66260_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0400011646.mm.1 | NM_025452.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400011646.mm.1 | NM_025452.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400011646.mm.1 | NM_133881.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400011646.mm.1 | NM_133881.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400011646.mm.1 | TC0400001512.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400011648.mm.1 | NM_025452.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400011648.mm.1 | NM_025452.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400011648.mm.1 | NM_133881.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400011648.mm.1 | NM_133881.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400011648.mm.1 | TC0400001512.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400011647.mm.1 | NM_133881.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400011647.mm.1 | NM_133881.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400011647.mm.1 | TC0400001512.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400011649.mm.1 | NM_025452.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400011649.mm.1 | NM_025452.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400011649.mm.1 | NM_133881.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400011649.mm.1 | NM_133881.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400011649.mm.1 | TC0400001512.mm.1 | MTA TC |
Affymetrix HT Mouse Genome 430A Array | 1417895_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417895_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 906092 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 780059 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 780059 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000780059 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1241297 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1241297 | - | - |
mouse-6 v1.1 (Illumina) | scl24923.6.1_23-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 780059 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl24923.6.1_23-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 780059 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2919393 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2919393 | - | - |
