Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027865.2 | NP_082141.1 | 7 - 8 | 997 | 59 | |
GenBank mRNA | AK002841.1 | - | 7 - 8 | 998 | 59 | |
AK090375.1 | - | 5 - 6 | 1271 | 59 | ||
AK154206.1 | - | 6 - 7 | 1483 | 59 | ||
AK165199.1 | - | 7 - 8 | 1029 | 59 | ||
AK165827.1 | - | 7 - 8 | 982 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |