Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028680.3 | NP_082956.2 | 8 - 9 | 1026 | 110 | |
GenBank mRNA | AF274590.1 | - | 8 - 9 | 1044 | 110 | |
AK014731.1 | - | 8 - 9 | 1078 | 110 | ||
AK040328.1 | - | 5 - 6 | 597 | 110 | ||
AK047217.1 | - | 8 - 9 | 1026 | 110 | ||
BC138143.1 | - | 8 - 9 | 1000 | 8861162 | 110 | |
BC138144.1 | - | 8 - 9 | 1000 | 8861163 | 110 | |
DQ266091.1 | - | 8 - 9 | 1067 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |