Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idh3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130884.4 | NP_570954.1 | 6 - 7 | 563 | 86 | |
GenBank mRNA | AK216020.1 | - | 1 - 2 | 101 | 86 | |
AY035212.1 | - | 6 - 7 | 544 | 86 | ||
BC009022.1 | - | 6 - 7 | 552 | 3257667 | 86 | |
CN456483.1 | - | 1 - 2 | 111 | 86 | ||
DN176034.1 | - | 6 - 7 | 529 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |