Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166456.1 | NP_001159928.1 | 4 - 5 | 616 | 64 | |
NM_001166458.1 | NP_001159930.1 | 4 - 5 | 823 | 64 | ||
NM_134086.4 | NP_598847.2 | 4 - 5 | 1309 | 64 | ||
GenBank mRNA | AF184240.2 | - | 3 - 4 | 401 | 64 | |
AK029189.1 | - | 4 - 5 | 486 | 64 | ||
AK034130.1 | - | 4 - 5 | 1311 | 64 | ||
AK035098.1 | - | 3 - 4 | 328 | 64 | ||
AK047459.1 | - | 4 - 5 | 840 | 64 | ||
AK049173.1 | - | 4 - 5 | 523 | 64 | ||
AK049294.1 | - | 4 - 5 | 589 | 64 | ||
AK050914.1 | - | 4 - 5 | 524 | 64 | ||
AK081724.1 | - | 4 - 5 | 825 | 64 | ||
AK082683.1 | - | 4 - 5 | 524 | 64 | ||
AK162612.1 | - | 4 - 5 | 674 | 64 | ||
AK165188.1 | - | 4 - 5 | 876 | 64 | ||
BC030378.1 | - | 4 - 5 | 525 | 5375319 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |