Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btnl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138678.2 | NP_619619.2 | 1 - 2 | 131 | 97 | |
GenBank mRNA | AF269232.1 | - | 1 - 2 | 131 | 97 | |
AK029784.1 | - | 1 - 2 | 237 | 97 | ||
AK143638.1 | - | 1 - 2 | 281 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |