Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019785.2 | NP_062759.2 | 7 - 8 | 691 | 85 | |
GenBank mRNA | AF190797.1 | - | 7 - 8 | 658 | 85 | |
AK090043.1 | - | 7 - 8 | 695 | 85 | ||
AK161642.1 | - | 7 - 8 | 651 | 85 | ||
BC002227.1 | - | 7 - 8 | 651 | 3491299 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |