Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aox4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023631.2 | NP_076120.2 | 18 - 19 | 2174 | 83 | |
GenBank mRNA | AF233581.1 | - | 18 - 19 | 2101 | 83 | |
AK158427.1 | - | 18 - 19 | 2175 | 83 | ||
AK160953.1 | - | 18 - 19 | 2140 | 83 | ||
BC117975.1 | - | 18 - 19 | 2101 | 40098789 | 83 | |
BC117976.1 | - | 18 - 19 | 2101 | 40098791 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |