Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aox4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023631.2 | NP_076120.2 | 26 - 27 | 3248 | 92 | |
GenBank mRNA | AF233581.1 | - | 26 - 27 | 3175 | 92 | |
AK009112.1 | - | 1 - 2 | 167 | 92 | ||
AK158427.1 | - | 26 - 27 | 3249 | 92 | ||
AK160953.1 | - | 26 - 27 | 3215 | 92 | ||
BC117975.1 | - | 26 - 27 | 3175 | 40098789 | 92 | |
BC117976.1 | - | 26 - 27 | 3175 | 40098791 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |