Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025442.3 | NP_079718.1 | 5 - 6 | 487 | 64 | |
GenBank mRNA | AK003818.1 | - | 5 - 6 | 468 | 64 | |
AK005296.1 | - | 5 - 6 | 488 | 64 | ||
AK006206.1 | - | 5 - 6 | 456 | 64 | ||
AK030567.1 | - | 5 - 6 | 458 | 64 | ||
AK161187.1 | - | 466 | 64 | |||
BC027160.1 | - | 5 - 6 | 447 | 4946430 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |