Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pop1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026340.3 | NP_080616.1 | 8 - 9 | 1369 | 64 | |
NM_152894.2 | NP_690854.1 | 8 - 9 | 1369 | 64 | ||
GenBank mRNA | AK016547.1 | - | 8 - 9 | 1356 | 64 | |
BC034742.1 | - | 8 - 9 | 1348 | 4935194 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |