Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dph5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027193.2 | NP_081469.2 | 1 - 2 | 95 | 76 | |
GenBank mRNA | AK010475.1 | - | 1 - 2 | 85 | 76 | |
AK034652.1 | - | 105 | 76 | |||
AK045127.1 | - | 1 - 2 | 115 | 76 | ||
AK045548.1 | - | 1 - 2 | 85 | 76 | ||
AK046024.1 | - | 1 - 2 | 85 | 76 | ||
AK046753.1 | - | 1 - 2 | 113 | 76 | ||
BC060372.1 | - | 1 - 2 | 95 | 6391546 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |