Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Katnal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027721.2 | NP_081997.1 | 7 - 8 | 723 | 86 | |
GenBank mRNA | AK014074.1 | - | 8 - 9 | 834 | 86 | |
AK017114.1 | - | 7 - 8 | 724 | 86 | ||
BC056963.1 | - | 7 - 8 | 670 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |