Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200006672.mm.1 |
NM_027990.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006672.mm.1 |
NM_027990.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006672.mm.1 |
TC0200000803.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006666.mm.1 |
NM_027990.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006666.mm.1 |
NM_027990.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006666.mm.1 |
TC0200000803.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006667.mm.1 |
NM_027990.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006667.mm.1 |
NM_027990.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006667.mm.1 |
TC0200000803.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006670.mm.1 |
NM_027990.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006670.mm.1 |
NM_027990.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006670.mm.1 |
TC0200000803.mm.1 |
MTA TC |