Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600010028.mm.1 |
TC0600001271.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600010028.mm.1 |
NM_001253717.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600010028.mm.1 |
NM_001253717.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600010028.mm.1 |
NM_001253718.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600010028.mm.1 |
NM_001253718.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600010028.mm.1 |
NM_028835.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600010028.mm.1 |
NM_028835.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600010021.mm.1 |
TC0600001271.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600010021.mm.1 |
NM_001253718.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600010021.mm.1 |
NM_001253718.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600010027.mm.1 |
TC0600001271.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600010027.mm.1 |
NM_001253717.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600010027.mm.1 |
NM_001253717.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600010027.mm.1 |
NM_001253718.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600010027.mm.1 |
NM_001253718.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600010027.mm.1 |
NM_028835.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600010027.mm.1 |
NM_028835.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600010025.mm.1 |
TC0600001271.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600010025.mm.1 |
NM_001253717.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600010025.mm.1 |
NM_001253717.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600010029.mm.1 |
TC0600001271.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600010029.mm.1 |
NM_001253717.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600010029.mm.1 |
NM_001253717.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600010029.mm.1 |
NM_001253718.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600010029.mm.1 |
NM_001253718.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600010029.mm.1 |
NM_028835.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600010029.mm.1 |
NM_028835.4 |
Refseq |