Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsun6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165941.1 | NP_001159413.1 | 8 - 9 | 1209 | 92 | |
NM_001165943.1 | NP_001159415.1 | 7 - 8 | 913 | 92 | ||
NM_028950.4 | NP_083226.3 | 7 - 8 | 1096 | 92 | ||
GenBank mRNA | AK016629.1 | - | 7 - 8 | 913 | 92 | |
AK016811.1 | - | 8 - 9 | 1222 | 92 | ||
AK077778.1 | - | 7 - 8 | 980 | 92 | ||
AK132428.1 | - | 7 - 8 | 1063 | 92 | ||
BC025622.1 | - | 4 - 5 | 439 | 92 | ||
BC053751.1 | - | 7 - 8 | 1064 | 6335896 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |