Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28046746 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28046742 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2120466 |
AK076332.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2120466 |
AK042409.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2120466 |
NM_028965.4 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2120466 |
NM_001163389.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2183383 |
AK076332.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2183383 |
BC014719.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2183383 |
AK042409.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2183383 |
NM_028965.4 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2183383 |
AK017118.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2183378 |
AK076332.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2183378 |
BC014719.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2183378 |
AK042409.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2183378 |
NM_028965.4 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2183378 |
AK017118.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
211727 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
70159 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
728389 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
599385 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
810748 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
446315 |
AK002794.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
446315 |
AK078399.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
446315 |
AK149557.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
446315 |
BC023117.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
446315 |
BC029637.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
446315 |
NM_144512.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
803542 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
107332 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P175298 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2183378 |
AK076332.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2183378 |
BC014719.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2183378 |
AK042409.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2183378 |
NM_028965.4 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2183378 |
AK017118.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2183383 |
AK076332.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2183383 |
BC014719.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2183383 |
AK042409.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2183383 |
NM_028965.4 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2183383 |
AK017118.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2120466 |
AK076332.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2120466 |
AK042409.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2120466 |
NM_028965.4 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2120466 |
NM_001163389.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28046764 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28046731 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28046762 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28046739 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28046728 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28046750 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28046743 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28046724 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28046770 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28046746 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28046755 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28046772 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28046722 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28046763 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28046752 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28046726 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28046742 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28046723 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28046757 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28046734 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28046737 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28046744 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28046748 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28046773 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28046741 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28046756 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28046725 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28046738 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28046767 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28046753 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07453843 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P175298 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P161429 |
AK076332.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P161429 |
BC014719.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P161429 |
AK042409.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P161429 |
NM_028965.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P161429 |
AK017118.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P253557 |
AK037747.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P116789 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P175298 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09651179 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09651176 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09651185 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09651173 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09456765 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09651177 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28046742 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28046746 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046660 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046667 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046713 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046654 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046706 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046739 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046688 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046772 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046716 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046659 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046650 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046658 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046673 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046690 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046665 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046696 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046770 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046719 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046686 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046737 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046735 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046693 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046647 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046675 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046741 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046679 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046682 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046645 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046728 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046684 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046757 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046743 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046746 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046725 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046723 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046766 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046702 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046680 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046764 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046670 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046705 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046692 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046712 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046668 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046724 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046756 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046671 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046726 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046649 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046648 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046709 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046685 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046694 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046677 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046663 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046731 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046652 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046695 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046759 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046715 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046744 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28046721 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09525098 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09651180 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046649 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046756 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046699 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046731 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09519602 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09651177 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046680 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09464527 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046667 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046660 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046668 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046670 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046751 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09519586 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046645 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046743 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046721 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046654 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046677 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046652 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046673 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046685 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09519597 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046658 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046686 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046650 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09525092 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046709 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046739 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09132875 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046713 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09651174 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09340823 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09609232 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046724 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046748 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09193462 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09651172 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09651179 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09525109 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046698 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046757 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046694 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09102747 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09672004 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046702 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046719 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046659 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046723 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046755 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09456765 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046706 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046700 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09075650 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046692 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046688 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046711 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046671 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046679 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046665 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28046744 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09519575 |
- |
- |