Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rprd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081293.1 | NP_001074762.1 | 2 - 3 | 537 | 113 | |
GenBank mRNA | AK012860.1 | - | 2 - 3 | 412 | 113 | |
AK015972.1 | - | 2 - 3 | 448 | 113 | ||
AK083674.1 | - | 2 - 3 | 360 | 113 | ||
AK087157.1 | - | 2 - 3 | 363 | 113 | ||
AK129150.1 | - | 2 - 3 | 216 | 113 | ||
AK154692.1 | - | 2 - 3 | 343 | 113 | ||
BC067054.1 | - | 2 - 3 | 342 | 30542928 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |