Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps33a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029929.3 | NP_084205.3 | 5 - 6 | 630 | 89 | |
GenBank mRNA | AF439858.1 | - | 5 - 6 | 614 | 89 | |
AK019463.1 | - | 5 - 6 | 614 | 89 | ||
AK041982.1 | - | 5 - 6 | 626 | 89 | ||
AK140553.1 | - | 5 - 6 | 619 | 89 | ||
AK150282.1 | - | 5 - 6 | 622 | 89 | ||
AK150550.1 | - | 5 - 6 | 622 | 89 | ||
BC046417.1 | - | 5 - 6 | 621 | 4501207 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |