Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tada1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030245.3 | NP_084521.1 | 1 - 2 | 312 | 92 | |
GenBank mRNA | AK050649.1 | - | 1 - 2 | 156 | 92 | |
AK151132.1 | - | 1 - 2 | 174 | 92 | ||
BC002307.1 | - | 1 - 2 | 114 | 3592046 | 92 | |
BC027337.1 | - | 1 - 2 | 179 | 3995355 | 92 | |
BY057637.1 | - | 1 - 2 | 313 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |