Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133981.2 | NP_598742.1 | 8 - 9 | 1054 | 115 | |
NR_045601.1 | 7 - 8 | 983 | 115 | |||
GenBank mRNA | AK050335.1 | - | 8 - 9 | 975 | 115 | |
AK054293.1 | - | 8 - 9 | 960 | 115 | ||
AK086674.1 | - | 8 - 9 | 994 | 115 | ||
AK149592.1 | - | 7 - 8 | 905 | 115 | ||
BC021791.1 | - | 8 - 9 | 949 | 4459738 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |