Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900009637.mm.1 |
NM_144533.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009637.mm.1 |
NM_144533.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009637.mm.1 |
TC0900001231.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900009648.mm.1 |
NM_144533.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009648.mm.1 |
NM_144533.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009648.mm.1 |
TC0900001231.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900009638.mm.1 |
NM_144533.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009638.mm.1 |
NM_144533.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009638.mm.1 |
TC0900001231.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900009639.mm.1 |
NM_144533.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009639.mm.1 |
NM_144533.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009639.mm.1 |
TC0900001231.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900009642.mm.1 |
NM_144533.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009642.mm.1 |
NM_144533.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009642.mm.1 |
TC0900001231.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900009647.mm.1 |
NM_144533.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009647.mm.1 |
NM_144533.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009647.mm.1 |
TC0900001231.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900009640.mm.1 |
NM_144533.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009640.mm.1 |
NM_144533.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009640.mm.1 |
TC0900001231.mm.1 |
MTA TC |