Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039520.2 | NP_001034609.1 | 6 - 7 | 1039 | 59 | |
NM_001253893.1 | NP_001240822.1 | 6 - 7 | 1039 | 59 | ||
NM_001253894.1 | NP_001240823.1 | 6 - 7 | 1039 | 59 | ||
NM_007871.2 | NP_031897.2 | 6 - 7 | 1039 | 59 | ||
GenBank mRNA | AK005012.2 | - | 6 - 7 | 1015 | 59 | |
AK015410.1 | - | 5 - 6 | 704 | 59 | ||
AK150046.1 | - | 6 - 7 | 1002 | 59 | ||
AK154375.1 | - | 5 - 6 | 703 | 59 | ||
AK157844.1 | - | 6 - 7 | 1009 | 59 | ||
AK170574.1 | - | 6 - 7 | 1015 | 59 | ||
AK171049.1 | - | 6 - 7 | 993 | 59 | ||
AK171566.1 | - | 6 - 7 | 1003 | 59 | ||
AK172231.1 | - | 6 - 7 | 1020 | 59 | ||
AK205128.1 | - | 1 - 2 | 41 | 59 | ||
L31398.1 | - | 6 - 7 | 979 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |