Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039520.2 | NP_001034609.1 | 12 - 13 | 1680 | 91 | |
NM_001253893.1 | NP_001240822.1 | 12 - 13 | 1680 | 91 | ||
NM_001253894.1 | NP_001240823.1 | 12 - 13 | 1680 | 91 | ||
NM_007871.2 | NP_031897.2 | 12 - 13 | 1680 | 91 | ||
GenBank mRNA | AK005012.2 | - | 12 - 13 | 1656 | 91 | |
AK015410.1 | - | 11 - 12 | 1345 | 91 | ||
AK150046.1 | - | 12 - 13 | 1643 | 91 | ||
AK157844.1 | - | 12 - 13 | 91 | |||
AK170574.1 | - | 12 - 13 | 1656 | 91 | ||
AK171049.1 | - | 12 - 13 | 1634 | 91 | ||
AK171566.1 | - | 12 - 13 | 1644 | 91 | ||
AK172231.1 | - | 12 - 13 | 1661 | 91 | ||
AK184894.1 | - | 2 - 3 | 142 | 91 | ||
AK191394.1 | - | 2 - 3 | 142 | 91 | ||
BC010233.1 | - | 3 - 4 | 396 | 3499181 | 91 | |
L31398.1 | - | 12 - 13 | 1620 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |