Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il18r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161842.1 | NP_001155314.1 | 2 - 3 | 422 | 67 | |
NM_008365.2 | NP_032391.1 | 1 - 2 | 347 | 67 | ||
GenBank mRNA | AK089237.1 | - | 2 - 3 | 423 | 67 | |
AK170705.1 | - | 2 - 3 | 383 | 67 | ||
U43673.1 | - | 1 - 2 | 347 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |