Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145779.1 | NP_001139251.1 | 18 - 19 | 1934 | 68 | |
NM_008442.2 | NP_032468.2 | 20 - 21 | 2431 | 68 | ||
GenBank mRNA | AK037037.1 | - | 18 - 19 | 1847 | 68 | |
BB622253.1 | - | 2 - 3 | 204 | 68 | ||
BC006803.1 | - | 18 - 19 | 1929 | 3152883 | 68 | |
D12644.1 | - | 20 - 21 | 2432 | 68 | ||
Y15894.1 | - | 17 - 18 | 1784 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |