Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039162.2 | NP_001034251.1 | 10 - 11 | 2838 | 63 | |
NM_009990.3 | NP_034120.2 | 11 - 12 | 2943 | 63 | ||
GenBank mRNA | AK045967.1 | - | 11 - 12 | 2788 | 63 | |
AK172928.1 | - | 11 - 12 | 2688 | 63 | ||
BC039162.1 | - | 10 - 11 | 2778 | 3498067 | 63 | |
BC053048.1 | - | 10 - 11 | 2838 | 5709279 | 63 | |
BC121191.1 | - | 10 - 11 | 2484 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |