Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dazl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277863.1 | NP_001264792.1 | 5 - 6 | 571 | 87 | |
NM_010021.5 | NP_034151.3 | 5 - 6 | 571 | 87 | ||
GenBank mRNA | AK014854.1 | - | 5 - 6 | 551 | 87 | |
AK033183.1 | - | 5 - 6 | 551 | 87 | ||
AK033316.1 | - | 5 - 6 | 551 | 87 | ||
AK160855.1 | - | 5 - 6 | 430 | 87 | ||
BC099940.1 | - | 5 - 6 | 531 | 30048233 | 87 | |
U46694.1 | - | 5 - 6 | 467 | 87 | ||
X95724.1 | - | 5 - 6 | 460 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |