Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Faah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010173.4 | NP_034303.3 | 1 - 2 | 442 | 76 | |
GenBank mRNA | AK004985.1 | - | 1 - 2 | 268 | 76 | |
AK032079.1 | - | 1 - 2 | 216 | 76 | ||
AK079598.1 | - | 1 - 2 | 443 | 76 | ||
AK080603.1 | - | 1 - 2 | 274 | 76 | ||
AK138481.1 | - | 1 - 2 | 62 | 76 | ||
BC052321.1 | - | 1 - 2 | 484 | 6515634 | 76 | |
U82536.1 | - | 1 - 2 | 203 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |