Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010180.2 | NP_034310.2 | 13 - 14 | 1700 | 60 | |
GenBank mRNA | AK035388.1 | - | 13 - 14 | 1653 | 60 | |
AK083573.1 | - | 13 - 14 | 1703 | 60 | ||
AK085989.1 | - | 13 - 14 | 1643 | 60 | ||
AK086451.1 | - | 13 - 14 | 1701 | 60 | ||
AK137511.1 | - | 13 - 14 | 1647 | 60 | ||
AK159646.1 | - | 13 - 14 | 1655 | 60 | ||
AK159816.1 | - | 2 - 3 | 145 | 60 | ||
BC007140.1 | - | 13 - 14 | 1644 | 3495754 | 60 | |
EU543224.1 | - | 13 - 14 | 1638 | 60 | ||
X70854.1 | - | 13 - 14 | 1697 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |