Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010266.2 | NP_034396.1 | 6 - 7 | 784 | 97 | |
GenBank mRNA | AF174583.1 | - | 6 - 7 | 710 | 97 | |
AF369880.1 | - | 6 - 7 | 669 | 97 | ||
AF369881.1 | - | 6 - 7 | 669 | 97 | ||
AF369882.1 | - | 6 - 7 | 669 | 97 | ||
AF369883.1 | - | 6 - 7 | 669 | 97 | ||
AF369884.1 | - | 6 - 7 | 669 | 97 | ||
AK041056.1 | - | 6 - 7 | 723 | 97 | ||
AK079419.1 | - | 6 - 7 | 592 | 97 | ||
AK079459.1 | - | 6 - 7 | 724 | 97 | ||
AK139617.1 | - | 551 | 97 | |||
AK157014.1 | - | 6 - 7 | 746 | 97 | ||
AK173138.1 | - | 6 - 7 | 713 | 97 | ||
BC076583.1 | - | 6 - 7 | 697 | 30620457 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |