Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012001.2 | NP_036131.1 | 3 - 4 | 477 | 88 | |
GenBank mRNA | AF071314.1 | - | 3 - 4 | 346 | 88 | |
AK003752.1 | - | 3 - 4 | 339 | 88 | ||
AK009955.1 | - | 3 - 4 | 349 | 88 | ||
AK162763.1 | - | 3 - 4 | 343 | 88 | ||
AK167833.1 | - | 3 - 4 | 363 | 88 | ||
BC116800.1 | - | 3 - 4 | 331 | 40126119 | 88 | |
BC116826.1 | - | 3 - 4 | 331 | 40126145 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |