Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012001.2 | NP_036131.1 | 7 - 8 | 1049 | 72 | |
GenBank mRNA | AF071314.1 | - | 7 - 8 | 918 | 72 | |
AK003752.1 | - | 7 - 8 | 916 | 72 | ||
AK009955.1 | - | 7 - 8 | 921 | 72 | ||
AK167833.1 | - | 7 - 8 | 935 | 72 | ||
BC020461.1 | - | 3 - 4 | 1471 | 72 | ||
BC116800.1 | - | 7 - 8 | 903 | 40126119 | 72 | |
BC116826.1 | - | 7 - 8 | 903 | 40126145 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |