Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012001.2 | NP_036131.1 | 8 - 9 | 1172 | 124 | |
GenBank mRNA | AF071314.1 | - | 8 - 9 | 1041 | 124 | |
AK003752.1 | - | 8 - 9 | 1039 | 124 | ||
AK009955.1 | - | 8 - 9 | 1044 | 124 | ||
AK167833.1 | - | 8 - 9 | 1058 | 124 | ||
BC020461.1 | - | 4 - 5 | 1594 | 124 | ||
BC116800.1 | - | 8 - 9 | 1026 | 40126119 | 124 | |
BC116826.1 | - | 8 - 9 | 1026 | 40126145 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |