Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013522.3 | NP_038550.2 | 3 - 4 | 413 | 97 | |
GenBank mRNA | AK038143.1 | - | 2 - 3 | 171 | 97 | |
AK077107.1 | - | 3 - 4 | 340 | 97 | ||
AK079229.1 | - | 3 - 4 | 305 | 97 | ||
BC002027.1 | - | 3 - 4 | 369 | 3487998 | 97 | |
CB573470.1 | - | 3 - 4 | 375 | 97 | ||
U62105.1 | - | 3 - 4 | 372 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |