Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013522.3 | NP_038550.2 | 6 - 7 | 698 | 116 | |
GenBank mRNA | AK038143.1 | - | 5 - 6 | 456 | 116 | |
AK077107.1 | - | 6 - 7 | 625 | 116 | ||
AK079229.1 | - | 6 - 7 | 590 | 116 | ||
AW496369.1 | - | 3 - 4 | 116 | |||
BC002027.1 | - | 6 - 7 | 654 | 3487998 | 116 | |
U62105.1 | - | 6 - 7 | 657 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |