Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013581.3 | NP_038609.3 | 8 - 9 | 2242 | 80 | |
GenBank mRNA | AF109377.2 | - | 8 - 9 | 2225 | 80 | |
AK031510.1 | - | 7 - 8 | 1660 | 80 | ||
AK034636.2 | - | 8 - 9 | 2247 | 80 | ||
AK152464.1 | - | 8 - 9 | 2233 | 80 | ||
AK158507.1 | - | 8 - 9 | 2604 | 80 | ||
AK159497.1 | - | 8 - 9 | 2244 | 80 | ||
AK162971.1 | - | 8 - 9 | 2243 | 80 | ||
AK171574.1 | - | 8 - 9 | 2236 | 80 | ||
AK220387.1 | - | 7 - 8 | 2686 | 80 | ||
BC049541.1 | - | 2 - 3 | 692 | 80 | ||
BC063056.1 | - | 8 - 9 | 2224 | 5720460 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |