Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600009061.mm.1 |
TC0600001176.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009061.mm.1 |
NM_017383.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009061.mm.1 |
NM_017383.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009055.mm.1 |
TC0600001176.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009055.mm.1 |
NM_017383.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009055.mm.1 |
NM_017383.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009059.mm.1 |
TC0600001176.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009059.mm.1 |
NM_017383.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009059.mm.1 |
NM_017383.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009062.mm.1 |
TC0600001176.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009062.mm.1 |
NM_017383.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009062.mm.1 |
NM_017383.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009056.mm.1 |
TC0600001176.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009056.mm.1 |
NM_017383.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009056.mm.1 |
NM_017383.3 |
Refseq |