Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017383.3 | NP_059079.2 | 6 - 7 | 970 | 71 | |
GenBank mRNA | AB032602.1 | - | 5 - 6 | 656 | 71 | |
AK052972.1 | - | 7 - 8 | 863 | 71 | ||
BC076594.1 | - | 6 - 7 | 970 | 30649661 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |