Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177371.1 | NP_001170842.1 | 7 - 8 | 831 | 62 | |
NM_001177372.1 | NP_001170843.1 | 7 - 8 | 831 | 62 | ||
NM_019813.4 | NP_062787.2 | 7 - 8 | 831 | 62 | ||
GenBank mRNA | AB064321.1 | - | 7 - 8 | 821 | 62 | |
AF187147.1 | - | 7 - 8 | 821 | 62 | ||
AF187148.1 | - | 7 - 8 | 821 | 62 | ||
AK132060.1 | - | 7 - 8 | 817 | 62 | ||
AK162697.1 | - | 7 - 8 | 818 | 62 | ||
BC006714.1 | - | 8 - 9 | 779 | 3710497 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |