Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chordc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025844.2 | NP_080120.1 | 9 - 10 | 913 | 102 | |
GenBank mRNA | AK003259.1 | - | 9 - 10 | 914 | 102 | |
AK132696.1 | - | 893 | 102 | |||
BC018374.1 | - | 9 - 10 | 913 | 4236535 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |