Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Knstrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026412.3 | NP_080688.2 | 1 - 2 | 307 | 89 | |
GenBank mRNA | AK006328.1 | - | 1 - 2 | 308 | 89 | |
AK012148.1 | - | 1 - 2 | 107 | 89 | ||
AK013184.1 | - | 1 - 2 | 56 | 89 | ||
AK028590.1 | - | 1 - 2 | 108 | 89 | ||
AK036374.1 | - | 1 - 2 | 96 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |