Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026538.3 | NP_080814.1 | 11 - 12 | 1396 | 74 | |
GenBank mRNA | AK011136.1 | - | 11 - 12 | 1406 | 74 | |
AK076047.1 | - | 11 - 12 | 1414 | 74 | ||
AK076975.1 | - | 11 - 12 | 1396 | 74 | ||
AK077514.1 | - | 11 - 12 | 1409 | 74 | ||
AK077953.1 | - | 6 - 7 | 643 | 74 | ||
AK145628.1 | - | 11 - 12 | 1437 | 74 | ||
AK159793.1 | - | 11 - 12 | 1400 | 74 | ||
AK169654.1 | - | 11 - 12 | 1411 | 74 | ||
BC018291.1 | - | 11 - 12 | 1390 | 4190167 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |