Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Herc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK017531.1 | - | 22 - 23 | 2733 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |